N-Boc-2-pyrroleboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
B4063-1G
B4063-5G
TCB4063-1GEA
66.36
CAD
TCB4063-1G
TCB4063-5G
N-Boc-2-pyrroleboronic acid (contains varying amounts of Anhydride)
N-Boc-2-pyrroleboronic acid
CAS Number: 135884-31-0
MDL Number: MFCD01318939
Molecular Formula: C9H14BNO4
Molecular Weight: 211.02
Form: Crystal
Color: White
Melting point (°C): 102
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1-Boc-2-pyrroleboronic Acid
Formula:
C9H14BNO4 Melting Pt: 102 °C |
MDL Number:
MFCD01318939 CAS Number: 135884-31-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...