N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane ≥90.0%

Supplier: TCI America

Synonyms: BCN-POE3-NH2, BCN-amine, rel-((1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-yl)methyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate, endo-BCN-PEG2-NH2

B4062-100MG B4062-25MG
TCB4062-100MGEA 1085.46 CAD
TCB4062-100MG TCB4062-25MG
N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane ≥90.0%
N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane

CAS Number: 1263166-93-3
MDL Number: MFCD19705418
Molecular Formula: C17H28N2O4
Molecular Weight: 324.42
Purity/Analysis Method: >90.0% (HPLC)
Form: Clear Liquid
Storage Temperature: >-20°C


Product Categories: Synthetic Organic Chemistry, Copper-free Click Reaction Reagents (Click Chemistry), Bioscience, Crosslinkers
Other Product Names / Synonyms: BCN-POE3-NH2, BCN-amine

Formula: C₁₇H₂₈N₂O₄
MDL Number: MFCD19705418
CAS Number: 1263166-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24959356b0a446becd83b3a1268af341660ed9f5&vid=24959356b0a446becd83b3a1268af341660ed9f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR