4-Bromo-1-methyl-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI America
B4060-25G
B4060-5G
TCB4060-25GEA
85.32
CAD
TCB4060-25G
TCB4060-5G
4-Bromo-1-methyl-1H-pyrazole ≥98.0% (by GC)
4-Bromo-1-methyl-1H-pyrazole
CAS Number: 15803-02-8
MDL Number: MFCD02179565
Molecular Formula: C4H5BrN2
Molecular Weight: 161.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 93
Specific Gravity (20/20): 1.61
Formula:
C₄H₅BrN₂ Boiling Pt: 78 °C Density: 1.61 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD02179565 CAS Number: 15803-02-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...