N,N-Bis(2-chloroethyl)-p-toluenesulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N,N-Bis(2-chloroethyl)-4-methylbenzenesulphonamide, N,N-Bis(2-chloroethyl)-4-methylbenzenesulfonamide

B4059-25G B4059-5G
TCB4059-25GEA 446.35 CAD
TCB4059-25G TCB4059-5G
N,N-Bis(2-chloroethyl)-p-toluenesulfonamide ≥98.0% (by HPLC, total nitrogen)
N,N-Bis(2-chloroethyl)-p-toluenesulfonamide

CAS Number: 42137-88-2
MDL Number: MFCD00018944
Molecular Formula: C11H15Cl2NO2S
Molecular Weight: 296.21
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 47

Formula: C₁₁H₁₅Cl₂NO₂S
MW: 296.21 g/mol
Melting Pt: 47 °C
MDL Number: MFCD00018944
CAS Number: 42137-88-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e385572649beeb676d54438075822ac038be78&vid=39e385572649beeb676d54438075822ac038be78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR