N-Benzyltropinone ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 8-Benzyl-8-azabicyclo[3.2.1]octan-3-one, (1R,5S)-8-Benzyl-8-azabicyclo[3.2.1]octan-3-one

B4058-1G B4058-5G
TCB4058-1GEA 50.56 CAD
TCB4058-1G TCB4058-5G
N-Benzyltropinone ≥95.0% (by GC, titration analysis)
N-Benzyltropinone

CAS Number: 28957-72-4
MDL Number: MFCD00151640
Molecular Formula: C14H17NO
Molecular Weight: 215.30
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 124
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: N-Benzyl-8-azabicyclo[3.2.1]octan-3-one

Formula: C₁₄H₁₇NO
MW: 215.30 g/mol
Boiling Pt: 124 °C
Density: 1.11 (20 °C)
MDL Number: MFCD00151640
CAS Number: 28957-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea16c2609789736fdb56011b74545bc9baeb569&vid=bea16c2609789736fdb56011b74545bc9baeb569&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR