2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(Bromomethyl)-2-(hydroxymethyl)propane-1,3-diol

B4057-1G B4057-5G
TCB4057-1GEA 121.66 CAD
TCB4057-1G TCB4057-5G
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol ≥98.0% (by GC)
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol

CAS Number: 19184-65-7
MDL Number: MFCD00135306
Molecular Formula: C5H11BrO3
Molecular Weight: 199.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77

Formula: C₅H₁₁BrO₃
MW: 199.04 g/mol
Melting Pt: 77 °C
MDL Number: MFCD00135306
CAS Number: 19184-65-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cef99a356dcb28f0440a1c7fe71723d2dc77c47&vid=7cef99a356dcb28f0440a1c7fe71723d2dc77c47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR