4-Bromo-2-methoxyphenol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B4056-25G B4056-5G
TCB4056-25GEA 530.88 CAD
TCB4056-25G TCB4056-5G
4-Bromo-2-methoxyphenol ≥97.0% (by GC, titration analysis)
4-Bromo-2-methoxyphenol

CAS Number: 7368-78-7
MDL Number: MFCD00051937
Molecular Formula: C7H7BrO2
Molecular Weight: 203.04
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 35


Other Product Names / Synonyms: 5-Bromo-2-hydroxyanisole, 4-Bromoguaiacol

Formula: C₇H₇BrO₂
Boiling Pt: 133 °C
Melting Pt: 35 °C
MDL Number: MFCD00051937
CAS Number: 7368-78-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56a50ca05d87aa4cc479af2447306ebfc4245a5a&vid=56a50ca05d87aa4cc479af2447306ebfc4245a5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR