4-(4-Bromophenylazo)phenol ≥98.0% (by GC)
Supplier: TCI America
B4053-1G
B4053-200MG
TCB4053-1GEA
1090.99
CAD
TCB4053-1G
TCB4053-200MG
4-(4-Bromophenylazo)phenol ≥98.0% (by GC)
4-(4-Bromophenylazo)phenol
CAS Number: 3035-94-7
MDL Number: MFCD00093929
Molecular Formula: C12H9BrN2O
Molecular Weight: 277.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 161
Lambda max.: 335 nm (THF)
Other Product Names / Synonyms: 4-Bromo-4'-hydroxyazobenzene
Formula:
C₁₂H₉BrN₂O Melting Pt: 161 °C |
MDL Number:
MFCD00093929 CAS Number: 3035-94-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...