3'-Bromo-2,2':5',2''-terthiophene ≥97.0%

Supplier: TCI America

Synonyms: 3-Bromo-2,5-di(2-thienyl)thiophene

B4050-1G B4050-5G
TCB4050-1GEA 685.72 CAD
TCB4050-1G TCB4050-5G
3'-Bromo-2,2':5',2''-terthiophene ≥97.0%
3'-Bromo-2,2':5',2''-terthiophene

CAS Number: 105125-00-6
Molecular Formula: C12H7BrS3
Molecular Weight: 327.27
Purity/Analysis Method: >97.0% (GC)
Melting point (°C): 40


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Bromo-2,5-di(2-thienyl)thiophene

Formula: C₁₂H₇BrS₃
Melting Pt: 40 °C
CAS Number: 105125-00-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b20accb933bb49f22db3cb15404a68e164c5c65&vid=8b20accb933bb49f22db3cb15404a68e164c5c65&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR