tert-Butyl (2-(methylamino)ethyl)carbamate ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-N'-methyl-1,2-diaminoethane, tert-Butyl N-(2-Methylaminoethyl)carbamate, N-(2-Methylaminoethyl)carbamic acid tert-Butyl ester, N-Boc-N'-methylethylenediamine, N-(tert-Butoxycarbonyl)-N'-methylethylenediamine, N-(tert-Butoxycarbonyl)-N'-methyl-1,2-diaminoethane

B4046-1G B4046-5G
TCB4046-1GEA 69.52 CAD
TCB4046-1G TCB4046-5G
tert-Butyl (2-(methylamino)ethyl)carbamate ≥98.0%
tert-Butyl (2-(methylamino)ethyl)carbamate

CAS Number: 122734-32-1
MDL Number: MFCD06808585
Molecular Formula: C8H18N2O2
Molecular Weight: 174.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Freezing point (°C): 35


Other Product Names / Synonyms: N-(tert-Butoxycarbonyl)-N'-methyl-1,2-diaminoethane, N-(2-Methylaminoethyl)carbamic Acid tert-Butyl Ester, N-Boc-N'-methyl-1,2-diaminoethane, N-Boc-N'-methylethylenediamine, tert-Butyl N-(2-Methylaminoethyl)carbamate

Formula: C₈H₁₈N₂O₂
MW: 174.2407 g/mol
MDL Number: MFCD06808585
CAS Number: 122734-32-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3172f993fbcd429c3ad84b77e4179a82f67e1333&vid=3172f993fbcd429c3ad84b77e4179a82f67e1333&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR