(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate ≥98.0%

Supplier: TCI America

Synonyms: Boc-β-iodo-Ala-OMe, BOC-3-iodo-L-alanine methyl ester, N-Boc-3-iodo-L-alanine Methyl Ester, Boc-β-Iodo-Ala-OMe, Boc-Ala(3-I)-OMe

B4044-1G B4044-5G
TCB4044-1GEA 69.36 CAD
TCB4044-1G TCB4044-5G
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate ≥98.0%
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate

CAS Number: 93267-04-0
MDL Number: MFCD00216579
Molecular Formula: C9H16INO4
Molecular Weight: 329.13
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 51
Specific rotation [a]20/D: -4 deg (C=3, MeOH)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Boc-3-iodo-L-alanine Methyl Ester, Boc-Ala(3-I)-OMe

Formula: C₉H₁₆INO₄
MW: 329.13 g/mol
Melting Pt: 51 °C
MDL Number: MFCD00216579
CAS Number: 93267-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8e74121e062b880cc0019bb6673bfb05b6ee596&vid=d8e74121e062b880cc0019bb6673bfb05b6ee596&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR