4-Bromopicolinic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromopyridine-2-carboxylic acid

B4041-1G B4041-5G
TCB4041-1GEA 52.14 CAD
TCB4041-1G TCB4041-5G
4-Bromopicolinic acid ≥98.0%
4-Bromopicolinic acid

CAS Number: 30766-03-1
MDL Number: MFCD03426665
Molecular Formula: C6H4BrNO2
Molecular Weight: 202.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 179


Other Product Names / Synonyms: 4-Bromopicolinic Acid

Formula: C₆H₄BrNO₂
Melting Pt: 179 °C
MDL Number: MFCD03426665
CAS Number: 30766-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd8b002feb048729c57135c3570798ccfa0742ed&vid=fd8b002feb048729c57135c3570798ccfa0742ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR