3-Bromotriphenylamine ≥98.0% (by GC)
Supplier: TCI America
B4036-1G
B4036-5G
TCB4036-1GEA
102.7
CAD
TCB4036-1G
TCB4036-5G
3-Bromotriphenylamine ≥98.0% (by GC)
3-Bromotriphenylamine
CAS Number: 78600-33-6
MDL Number: MFCD06798123
Molecular Formula: C18H14BrN
Molecular Weight: 324.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 193
Melting point (°C): 92
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Bromo-N,N-diphenylaniline
Formula:
C₁₈H₁₄BrN Boiling Pt: 193 °C Melting Pt: 92 °C |
MDL Number:
MFCD06798123 CAS Number: 78600-33-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...