1-Bromo-4-(phenylethynyl)benzene ≥98.0%

Supplier: TCI America

Synonyms: 1-(4-Bromophenyl)-2-phenylacetylene

B4024-1G B4024-5G
TCB4024-1GEA 107.44 CAD
TCB4024-1G TCB4024-5G
1-Bromo-4-(phenylethynyl)benzene ≥98.0%
1-Bromo-4-(phenylethynyl)benzene

CAS Number: 13667-12-4
MDL Number: MFCD01596910
Molecular Formula: C14H9Br
Molecular Weight: 257.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85


Other Product Names / Synonyms: 1-(4-Bromophenyl)-2-phenylacetylene

Formula: C₁₄H₉Br
Melting Pt: 85 °C
MDL Number: MFCD01596910
CAS Number: 13667-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1565a6d0c6392e5ad728bafade427457dd64fce&vid=c1565a6d0c6392e5ad728bafade427457dd64fce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR