1,4-Bis[(1H-imidazol-1-yl)methyl]benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: alpha,alpha'-Bis[(1-imidazolyl)methyl]-p-xylene, 1,4-Bis((1H-imidazol-1-yl)methyl)benzene

B4023-5G B4023-1G
TCB4023-5GEA 450.3 CAD
TCB4023-5G 76219-368
1,4-Bis[(1H-imidazol-1-yl)methyl]benzene ≥98.0% (by GC)
1,4-Bis[(1H-imidazol-1-yl)methyl]benzene

CAS Number: 56643-83-5
MDL Number: MFCD06598031
Molecular Formula: C14H14N4
Molecular Weight: 238.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 132


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: alpha,alpha'-Bis[(1-imidazolyl)methyl]-p-xylene

Formula: C₁₄H₁₄N₄
MW: 238.29 g/mol
Melting Pt: 132 °C
MDL Number: MFCD06598031
CAS Number: 56643-83-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d25341bc70a8076b1d319b5876dead9ea43fe956&vid=d25341bc70a8076b1d319b5876dead9ea43fe956&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR