4-Bromo-3-fluorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

B4022-25G B4022-5G
TCB4022-25GEA 347.6 CAD
TCB4022-25G TCB4022-5G
4-Bromo-3-fluorobenzonitrile ≥98.0% (by GC)
4-Bromo-3-fluorobenzonitrile

CAS Number: 133059-44-6
MDL Number: MFCD07782069
Molecular Formula: C7H3BrFN
Molecular Weight: 200.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 94

Formula: C₇H₃BrFN
MW: 200.01 g/mol
Melting Pt: 94 °C
MDL Number: MFCD07782069
CAS Number: 133059-44-6
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=776aad08a51e0938851e45ffe06bf295e209c351&vid=776aad08a51e0938851e45ffe06bf295e209c351&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR