2-Bromo-6-fluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4017-1G B4017-5G
TCB4017-1GEA 61.15 CAD
TCB4017-1G TCB4017-5G
2-Bromo-6-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-6-fluorobenzoic acid

CAS Number: 2252-37-1
MDL Number: MFCD01569539
Molecular Formula: C7H4BrFO2
Molecular Weight: 219.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 156

Formula: C₇H₄BrFO₂
Melting Pt: 156 °C
MDL Number: MFCD01569539
CAS Number: 2252-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abd325ec0fb7bf47597ccc1f2fc8060981849a01&vid=abd325ec0fb7bf47597ccc1f2fc8060981849a01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR