3-(tert-Butyl)-2-hydroxybenzaldehyde ≥95.0%

Supplier: TCI America

Synonyms: 3-tert-Butylsalicylaldehyde, 3-tert-Butyl-2-hydroxybenzaldehyde

B4016-1G B4016-5G
TCB4016-1GEA 61.62 CAD
TCB4016-1G TCB4016-5G
3-(tert-Butyl)-2-hydroxybenzaldehyde ≥95.0%
3-(tert-Butyl)-2-hydroxybenzaldehyde

CAS Number: 24623-65-2
MDL Number: MFCD00778877
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 113
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 3-tert-Butyl-2-hydroxybenzaldehyde

Formula: C11H14O2
Boiling Pt: 79 °C
Density: 1.05 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00778877
CAS Number: 24623-65-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20c151e75111dff04eca147caac603bf4f99bd4f&vid=20c151e75111dff04eca147caac603bf4f99bd4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR