1-Boc-4-(methylamino)piperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(Methylamino)piperidine, N1-BOC protected, tert-Butyl 4-(methylamino)piperidine-1-carboxylate

B4015-1G B4015-5G
TCB4015-1GEA 48.98 CAD
TCB4015-1G TCB4015-5G
1-Boc-4-(methylamino)piperidine ≥98.0% (by GC, titration analysis)
1-Boc-4-(methylamino)piperidine

CAS Number: 147539-41-1
MDL Number: MFCD02259411
Molecular Formula: C11H22N2O2
Molecular Weight: 214.31
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.00
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Boc-4-(methylamino)piperidine, tert-Butyl 4-Methylamino-1-piperidinecarboxylate, 4-Methylamino-1-piperidinecarboxylic Acid tert-Butyl Ester

Formula: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
Density: 1.00 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD02259411
CAS Number: 147539-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36fd1fcfb2153042879b95e3727ca06b7d998922&vid=36fd1fcfb2153042879b95e3727ca06b7d998922&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR