(±)-Boc-α-phosphonoglycine trimethyl ester ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate
CAS Number: 89524-98-1
MDL Number: MFCD02178893
Molecular Formula: C10H20NO7P
Molecular Weight: 297.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 71
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Trimethyl 2-(tert-Butoxycarbonylamino)-2-phosphonoacetate, Trimethyl 2-(Boc-amino)-2-phosphonoacetate, N-Boc-2-phosphonoglycine Trimethyl Ester
Formula:
C₁₀H₂₀NO₇P MW: 297.24 g/mol Melting Pt: 71 °C |
MDL Number:
MFCD02178893 CAS Number: 89524-98-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...