(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: GSK 3 Inhibitor IX, BIO, 6BIO, 6-Bromoindirubin-3'-oxime, (2Z,3E)-6'-Bromo-3-(hydroxyimino)-[2,3'-biindolinylidene]-2'-one

B4006-25MG B4006-5MG
TCB4006-25MGEA 568.8 CAD
TCB4006-25MG TCB4006-5MG
(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥95.0% (by HPLC)
(2'Z,3'E)-6-Bromoindirubin-3'-oxime

CAS Number: 667463-62-9
MDL Number: MFCD08705318
Molecular Formula: C16H10BrN3O2
Molecular Weight: 356.18
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 300
Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents
Other Product Names / Synonyms: BIO

Formula: C₁₆H₁₀BrN₃O₂
MW: 356.18 g/mol
Melting Pt: 300 °C
MDL Number: MFCD08705318
CAS Number: 667463-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d48b7fe92a87336e018d8fa7dc3bacfe36417ed5&vid=d48b7fe92a87336e018d8fa7dc3bacfe36417ed5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR