1-Boc-1-methylhydrazine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B3998-25G B3998-5G
TCB3998-25GEA 349.18 CAD
TCB3998-25G TCB3998-5G
1-Boc-1-methylhydrazine ≥97.0% (by GC, titration analysis)
1-Boc-1-methylhydrazine

CAS Number: 21075-83-2
MDL Number: MFCD05669700
Molecular Formula: C6H14N2O2
Molecular Weight: 146.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 71
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: 1-tert-Butoxycarbonyl-1-methylhydrazine, 1-Boc-1-methylhydrazine, 2-Methylcarbazic Acid tert-Butyl Ester

Formula: C₆H₁₄N₂O₂
Boiling Pt: 80 °C
Density: 0.99 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD05669700
CAS Number: 21075-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e82a777a9a2f19367340a9f553d77b8a2f691a35&vid=e82a777a9a2f19367340a9f553d77b8a2f691a35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR