2-Bromo-5-hydroxybenzaldehyde ≥97.0% (by GC)

Supplier: TCI America

B3997-1G B3997-5G
TCB3997-1GEA 48.98 CAD
TCB3997-1G TCB3997-5G
2-Bromo-5-hydroxybenzaldehyde ≥97.0% (by GC)
2-Bromo-5-hydroxybenzaldehyde

CAS Number: 2973-80-0
MDL Number: MFCD03033065
Molecular Formula: C7H5BrO2
Molecular Weight: 201.02
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 132

Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 132 °C
MDL Number: MFCD03033065
CAS Number: 2973-80-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4889793a24ebe0904868963f8d970d7f689660a&vid=d4889793a24ebe0904868963f8d970d7f689660a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR