Bis(4-bromophenyl)acetylene ≥98.0% (by GC)

Supplier: TCI America

B3996-1G B3996-5G
TCB3996-1GEA 95.91 CAD
TCB3996-1G TCB3996-5G
Bis(4-bromophenyl)acetylene ≥98.0% (by GC)
Bis(4-bromophenyl)acetylene

CAS Number: 2789-89-1
MDL Number: MFCD00185147
Molecular Formula: C14H8Br2
Molecular Weight: 336.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 187

Formula: C₁₄H₈Br₂
Melting Pt: 187 °C
MDL Number: MFCD00185147
CAS Number: 2789-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c3f79d8afea6958dc236486d2939fc6485801d9&vid=2c3f79d8afea6958dc236486d2939fc6485801d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR