(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3995-1G B3995-5G
TCB3995-1GEA 74.26 CAD
TCB3995-1G TCB3995-5G
(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid ≥98.0% (by HPLC, titration analysis)
(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid

CAS Number: 124655-17-0
MDL Number: MFCD00065575
Molecular Formula: C11H21NO4
Molecular Weight: 231.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 120
Specific rotation [a]20/D: 5 deg (C=1, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-tert-leucine, Boc-D-Tle-OH, (R)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid, (R)-2-(Boc-amino)-3,3-dimethylbutyric Acid

Formula: C₁₁H₂₁NO₄
Melting Pt: 120 °C
MDL Number: MFCD00065575
CAS Number: 124655-17-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=419d728a53549166aa53b1b788cdef1d4365ff7f&vid=419d728a53549166aa53b1b788cdef1d4365ff7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR