tert-Butyl-3-oxoazetidine-1-carboxylate ≥98.0% (by GC)

Supplier: TCI America

B3988-1G B3988-5G
TCB3988-1GEA 80.58 CAD
TCB3988-1G TCB3988-5G
tert-Butyl-3-oxoazetidine-1-carboxylate ≥98.0% (by GC)
tert-Butyl-3-oxoazetidine-1-carboxylate

CAS Number: 398489-26-4
MDL Number: MFCD01861741
Molecular Formula: C8H13NO3
Molecular Weight: 171.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 52
Flash Point (°C): 101


Other Product Names / Synonyms: 1-Boc-3-azetidinone, tert-Butyl 3-Oxoazetidine-1-carboxylate, 3-Oxoazetidine-1-carboxylic Acid tert-Butyl Ester

Formula: C₈H₁₃NO₃
Melting Pt: 52 °C
Flash Pt: 101 °C
MDL Number: MFCD01861741
CAS Number: 398489-26-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=369f0ab848191ff40a06d3ccb18df2e30955853a&vid=369f0ab848191ff40a06d3ccb18df2e30955853a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR