(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-tert-Butyl 2-Methyl (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-Butyl 2-Methyl Ester, N-Boc-cis-4-hydroxy-L-proline Methyl Ester, Boc-cis-Hyp-OMe

B3987-1G B3987-5G
TCB3987-1GEA 55.3 CAD
TCB3987-1G TCB3987-5G
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥97.0% (by GC)
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate

CAS Number: 102195-79-9
MDL Number: MFCD00237541
Molecular Formula: C11H19NO5
Molecular Weight: 245.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 82
Specific rotation [a]20/D: -64 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Boc-cis-Hyp-OMe, N-Boc-cis-4-hydroxy-L-proline Methyl Ester, (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-Butyl 2-Methyl Ester, 1-tert-Butyl 2-Methyl (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylate

Formula: C11H19NO5
MW: 245.28 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00237541
CAS Number: 102195-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96599fb785ceb3f40607b46e2d02405688cdcf80&vid=96599fb785ceb3f40607b46e2d02405688cdcf80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR