3,3'-Bipyridyl ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3984-1G B3984-5G
TCB3984-1GEA 246.48 CAD
TCB3984-1G TCB3984-5G
3,3'-Bipyridyl ≥98.0% (by GC, titration analysis)
3,3'-Bipyridyl

CAS Number: 581-46-4
MDL Number: MFCD00006371
Molecular Formula: C10H8N2
Molecular Weight: 156.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 175
Specific Gravity (20/20): 1.16


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 3,3'-Bipyridine, 3,3'-Dipyridyl

Formula: C₁₀H₈N₂
Boiling Pt: 175 °C
Density: 1.16 (20 °C)
MDL Number: MFCD00006371
CAS Number: 581-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b30876e031d0c2febe0727188d5268091b5c154&vid=8b30876e031d0c2febe0727188d5268091b5c154&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR