4,4'-Bipiperidine ≥98.0%
Supplier: TCI America
B3983-1G
B3983-5G
TCB3983-1GEA
157.51
CAD
TCB3983-1G
TCB3983-5G
4,4'-Bipiperidine ≥98.0%
4,4'-Bipiperidine
CAS Number: 15336-72-8
MDL Number: MFCD01104315
Molecular Formula: C10H20N2
Molecular Weight: 168.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 171
Formula:
C₁₀H₂₀N₂ Melting Pt: 171 °C |
MDL Number:
MFCD01104315 CAS Number: 15336-72-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...