1-(4-Bromophenyl)naphthalene ≥98.0% (by GC)

Supplier: TCI America

B3978-1G B3978-5G
TCB3978-1GEA 54.51 CAD
TCB3978-1G TCB3978-5G
1-(4-Bromophenyl)naphthalene ≥98.0% (by GC)
1-(4-Bromophenyl)naphthalene

CAS Number: 204530-94-9
MDL Number: MFCD07369916
Molecular Formula: C16H11Br
Molecular Weight: 283.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Formula: C₁₆H₁₁Br
MDL Number: MFCD07369916
CAS Number: 204530-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b346dbce35097700f64d7e6eba35bc001f8ece7b&vid=b346dbce35097700f64d7e6eba35bc001f8ece7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR