9-Bromo-10-phenylanthracene ≥98.0% (by GC)

Supplier: TCI America

B3977-1G B3977-5G
TCB3977-1GEA 44.24 CAD
TCB3977-1G TCB3977-5G
9-Bromo-10-phenylanthracene ≥98.0% (by GC)
9-Bromo-10-phenylanthracene

CAS Number: 23674-20-6
MDL Number: MFCD00230983
Molecular Formula: C20H13Br
Molecular Weight: 333.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 156

Formula: C₂₀H₁₃Br
Melting Pt: 156 °C
MDL Number: MFCD00230983
CAS Number: 23674-20-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37e8da9db082f8433b1b1dce4c54c9664250ae08&vid=37e8da9db082f8433b1b1dce4c54c9664250ae08&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR