Boc-glycine methyl ester ≥98.0%
Supplier: TCI America
Synonyms:
N-Boc-glycine Methyl Ester, Boc-Gly-OMe
B3975-25G
B3975-5G
TCB3975-25GEA
135.88
CAD
TCB3975-25G
TCB3975-5G
Boc-glycine methyl ester ≥98.0%
Boc-glycine methyl ester
CAS Number: 31954-27-5
MDL Number: MFCD00038267
Molecular Formula: C8H15NO4
Molecular Weight: 189.21
Purity/Analysis Method: >98.0% (N)
Form: Clear Liquid
Flash Point (°C): 109
Specific Gravity (20/20): 1.08
Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Boc-glycine Methyl Ester, Boc-Gly-OMe
Formula:
C8H15NO4 MW: 189.2 g/mol Density: 1.08 (20 °C) Flash Pt: 109 °C |
MDL Number:
MFCD00038267 CAS Number: 31954-27-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...