7-Bromoisatin ≥97.0%
Supplier: TCI America
Synonyms:
7-Bromo-1H-indole-2,3-dione, 7-Bromoindoline-2,3-dione
B3974-1G
B3974-5G
TCB3974-1GEA
34.76
CAD
TCB3974-1G
TCB3974-5G
7-Bromoisatin ≥97.0%
7-Bromoisatin
CAS Number: 20780-74-9
MDL Number: MFCD00774354
Molecular Formula: C8H4BrNO2
Molecular Weight: 226.03
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 200
Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 7-Bromoindole-2,3-dione, 7-Bromo-2,3-indolinedione
Formula:
C₈H₄BrNO₂ MW: 226.03 Melting Pt: 200 °C |
MDL Number:
MFCD00774354 CAS Number: 20780-74-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...