1,2-Bis(4'-methyl-2,2'-bipyridin-4-yl)ethane ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 4,4''-(1,2-Ethanediyl)bis(4'-methyl-2,2'-bipyridine)

B3973-1G B3973-200MG
TCB3973-1GEA 469.26 CAD
TCB3973-1G TCB3973-200MG
1,2-Bis(4'-methyl-2,2'-bipyridin-4-yl)ethane ≥96.0% (by GC)
1,2-Bis(4'-methyl-2,2'-bipyridin-4-yl)ethane

CAS Number: 96897-04-0
Molecular Formula: C24H22N4
Molecular Weight: 366.47
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 192


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 4,4''-(1,2-Ethanediyl)bis(4'-methyl-2,2'-bipyridine)

Formula: C₂₄H₂₂N₄
MW: 366.47 g/mol
Melting Pt: 192 °C
CAS Number: 96897-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ad53861d9865d4cb0280cbdb27d5e30b1297a55&vid=5ad53861d9865d4cb0280cbdb27d5e30b1297a55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR