tert-Butyl pyrrolidine-1-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: tert-butyl pyrrolidine-1-carboxylate, pyrrolidine-1-carboxylic acid tert-butyl ester, N-BOC-Pyrrolidine

B3971-25G B3971-5G
TCB3971-25GEA 315.68 CAD
TCB3971-25G TCB3971-5G
tert-Butyl pyrrolidine-1-carboxylate ≥98.0%
tert-Butyl pyrrolidine-1-carboxylate

CAS Number: 86953-79-9
MDL Number: MFCD00216581
Molecular Formula: C9H17NO2
Molecular Weight: 171.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 75
Flash Point (°C): 86
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: 1-Boc-pyrrolidine, 1-Pyrrolidinecarboxylic Acid tert-Butyl Ester, tert-Butyl 1-Pyrrolidinecarboxylate

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Boiling Pt: 75 °C
Density: 0.98 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00216581
CAS Number: 86953-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef83b9a7d48bb8c0e20d339b028440baaf8ba2fe&vid=ef83b9a7d48bb8c0e20d339b028440baaf8ba2fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR