3-Bromo-5-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Bromo-3-pyridinol, 3-Bromo-5-pyridinol, 5-Bromo-pyridin-3-ol

B3969-1G B3969-5G
TCB3969-1GEA 53.72 CAD
TCB3969-1G TCB3969-5G
3-Bromo-5-hydroxypyridine ≥98.0% (by GC, titration analysis)
3-Bromo-5-hydroxypyridine

CAS Number: 74115-13-2
MDL Number: MFCD00661305
Molecular Formula: C5H4BrNO
Molecular Weight: 174.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 169


Other Product Names / Synonyms: 5-Bromo-3-pyridinol

Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 169 °C
MDL Number: MFCD00661305
CAS Number: 74115-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07b23090fc587224bcf7893f41e321e193471e78&vid=07b23090fc587224bcf7893f41e321e193471e78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR