5-Bromo-2-chloro-3-nitropyridine ≥98.0%

Supplier: TCI America

B3963-25G B3963-5G
TCB3963-25GEA 231.47 CAD
TCB3963-25G TCB3963-5G
5-Bromo-2-chloro-3-nitropyridine ≥98.0%
5-Bromo-2-chloro-3-nitropyridine

CAS Number: 67443-38-3
MDL Number: MFCD00222270
Molecular Formula: C5H2BrClN2O2
Molecular Weight: 237.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68

Formula: C₅H₂BrClN₂O₂
MW: 237.44 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00222270
CAS Number: 67443-38-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03fb3146252c95f7229fe14e1a03ca4ad27e6f53&vid=03fb3146252c95f7229fe14e1a03ca4ad27e6f53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR