(2-Bromoethyl)diphenylsulfonium trifluoromethanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: (2-Bromoethyl)diphenylsulfonium Triflate, (2-Bromoethyl)diphenylsulphonium Triflate

B3958-1G B3958-5G
TCB3958-1GEA 83.74 CAD
TCB3958-1G TCB3958-5G
(2-Bromoethyl)diphenylsulfonium trifluoromethanesulfonate ≥98.0%
(2-Bromoethyl)diphenylsulfonium trifluoromethanesulfonate

CAS Number: 247129-85-7
MDL Number: MFCD12546049
Molecular Formula: C15H14BrF3O3S2
Molecular Weight: 443.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 90


Other Product Names / Synonyms: (2-Bromoethyl)diphenylsulfonium Triflate

Formula: C₁₅H₁₄BrF₃O₃S₂
Melting Pt: 90 °C
MDL Number: MFCD12546049
CAS Number: 247129-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ef32f86f8ec46bd032899678c3586e64601048a&vid=9ef32f86f8ec46bd032899678c3586e64601048a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR