2-Bromo-7-tert-butylpyrene ≥98.0% (by GC)

Supplier: TCI America

B3957-1G B3957-5G
TCB3957-1GEA 455.04 CAD
TCB3957-1G TCB3957-5G
2-Bromo-7-tert-butylpyrene ≥98.0% (by GC)
2-Bromo-7-tert-butylpyrene

CAS Number: 78751-80-1
Molecular Formula: C20H17Br
Molecular Weight: 337.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 152

Formula: C₂₀H₁₇Br
MW: 337.26 g/mol
Melting Pt: 152 °C
CAS Number: 78751-80-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d69bb53dc0a79c2d639e807031e11d5583612a57&vid=d69bb53dc0a79c2d639e807031e11d5583612a57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR