2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene ≥97.0%
Supplier: TCI America
Synonyms:
Pyrene-2,7-diboronic acid, pinacol ester
B3956-5G
B3956-1G
TCB3956-5GEA
1224.5
CAD
TCB3956-5G
TCB3956-1G
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene ≥97.0%
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene
CAS Number: 688756-58-3
MDL Number: MFCD15143601
Molecular Formula: C28H32B2O4
Molecular Weight: 454.18
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Lambda max.: 324 nm (EtOH)
Formula:
C₂₈H₃₂B₂O₄ MW: 454.18 g/mol |
MDL Number:
MFCD15143601 CAS Number: 688756-58-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...