9-Bromotriptycene ≥98.0% (by GC)

Supplier: TCI America

B3955-1G B3955-5G
TCB3955-1GEA 222.78 CAD
TCB3955-1G TCB3955-5G
9-Bromotriptycene ≥98.0% (by GC)
9-Bromotriptycene

CAS Number: 15364-55-3
MDL Number: MFCD01099022
Molecular Formula: C20H13Br
Molecular Weight: 333.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 254


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 9-Bromo-9,10-dihydro-9,10-[1,2]benzenoanthracene

Formula: C₂₀H₁₃Br
Melting Pt: 254 °C
MDL Number: MFCD01099022
CAS Number: 15364-55-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3596de3fb590afc2e811f3e05285acbcac1540b6&vid=3596de3fb590afc2e811f3e05285acbcac1540b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR