(2-Bromophenyl)diphenylphosphine ≥95.0% (by GC)

Supplier: TCI America

B3953-1G B3953-5G
TCB3953-1GEA 154.84 CAD
TCB3953-1G TCB3953-5G
(2-Bromophenyl)diphenylphosphine ≥95.0% (by GC)
(2-Bromophenyl)diphenylphosphine

CAS Number: 62336-24-7
MDL Number: MFCD00045014
Molecular Formula: C18H14BrP
Molecular Weight: 341.19
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 112

Formula: C₁₈H₁₄BrP
Melting Pt: 112 °C
MDL Number: MFCD00045014
CAS Number: 62336-24-7

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR