4-Bromo-N-phenylaniline ≥98.0% (by GC)

Supplier: TCI America

B3949-25G B3949-5G
TCB3949-25GEA 233.84 CAD
TCB3949-25G TCB3949-5G
4-Bromo-N-phenylaniline ≥98.0% (by GC)
4-Bromo-N-phenylaniline

CAS Number: 54446-36-5
MDL Number: MFCD07779504
Molecular Formula: C12H10BrN
Molecular Weight: 248.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 318
Melting point (°C): 87


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: N-(4-Bromophenyl)aniline

Formula: C₁₂H₁₀BrN
MW: 248.1200 g/mol
Boiling Pt: 318 °C
Melting Pt: 87 °C
MDL Number: MFCD07779504
CAS Number: 54446-36-5

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR