4-Bromo-2-chlorobenzaldehyde ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2-Chloro-4-bromobenzaldehyde

B3948-1G B3948-5G
TCB3948-1GEA 36.34 CAD
TCB3948-1G TCB3948-5G
4-Bromo-2-chlorobenzaldehyde ≥97.0% (by GC)
4-Bromo-2-chlorobenzaldehyde

CAS Number: 158435-41-7
MDL Number: MFCD08741413
Molecular Formula: C7H4BrClO
Molecular Weight: 219.46
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 75

Formula: C₇H₄BrClO
MW: 219.46 g/mol
Melting Pt: 75 °C
MDL Number: MFCD08741413
CAS Number: 158435-41-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e18c41b2ff67160783ca2c8d50aa93e91c4f7e2&vid=6e18c41b2ff67160783ca2c8d50aa93e91c4f7e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR