5-Bromo-2-chlorobenzyl alcohol ≥98.0% (by GC)
Supplier: TCI America
B3946-1G
B3946-5G
TCB3946-1GEA
95.91
CAD
TCB3946-1G
TCB3946-5G
5-Bromo-2-chlorobenzyl alcohol ≥98.0% (by GC)
5-Bromo-2-chlorobenzyl alcohol
CAS Number: 149965-40-2
MDL Number: MFCD02683548
Molecular Formula: C7H6BrClO
Molecular Weight: 221.48
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 94
Formula:
C₇H₆BrClO Melting Pt: 94 °C |
MDL Number:
MFCD02683548 CAS Number: 149965-40-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...