N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: Boc-D-Phe(4-Br)-OH, N-Boc-4-bromo-D-phenylalanine

B3945-1G B3945-5G
TCB3945-1GEA 53.72 CAD
TCB3945-1G TCB3945-5G
N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine ≥98.0%
N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine

CAS Number: 79561-82-3
MDL Number: MFCD00270359
Molecular Formula: C14H18BrNO4
Molecular Weight: 344.21
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 119
Specific rotation [a]20/D: -14 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Boc-D-Phe(4-Br)-OH, N-Boc-4-bromo-D-phenylalanine

Formula: C₁₄H₁₈BrNO₄
MW: 344.21 g/mol
Melting Pt: 119 °C
MDL Number: MFCD00270359
CAS Number: 79561-82-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e08d0bfbd9588f1da3bb1fb4c7fd1356333e7f6b&vid=e08d0bfbd9588f1da3bb1fb4c7fd1356333e7f6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR