2-Bromobenzophenone ≥97.0% (by GC)

Supplier: TCI America

B3937-1G B3937-5G
TCB3937-1GEA 34.76 CAD
TCB3937-1G TCB3937-5G
2-Bromobenzophenone ≥97.0% (by GC)
2-Bromobenzophenone

CAS Number: 13047-06-8
MDL Number: MFCD00032462
Molecular Formula: C13H9BrO
Molecular Weight: 261.12
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 163
Melting point (°C): 42

Formula: C₁₃H₉BrO
Boiling Pt: 163 °C
Melting Pt: 42 °C
MDL Number: MFCD00032462
CAS Number: 13047-06-8
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15e7525fc95dfaee4f6bc2b3caad350804d3e10f&vid=15e7525fc95dfaee4f6bc2b3caad350804d3e10f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR