4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0%

Supplier: TCI America

Synonyms: 4-tert-Butyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-tert-Butyl Ester, N-Boc-L-aspartic Acid 4-tert-Butyl Ester, Boc-Asp(OtBu)-OH

B3935-1G B3935-5G
TCB3935-1GEA 42.66 CAD
TCB3935-1G TCB3935-5G
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0%
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate

CAS Number: 1676-90-0
MDL Number: MFCD00076912
Molecular Formula: C13H23NO6
Molecular Weight: 289.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 63
Specific rotation [a]20/D: 2 deg (C=1, MeOH)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-tert-Butyl Ester, 4-tert-Butyl N-Boc-L-aspartate, N-Boc-L-aspartic Acid 4-tert-Butyl Ester, Boc-Asp(OtBu)-OH

Formula: C₁₃H₂₃NO₆
MW: 289.33 g/mol
Melting Pt: 63 °C
MDL Number: MFCD00076912
CAS Number: 1676-90-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4390bd3e3d1ec8b1216e4deba98e50160f4c480a&vid=4390bd3e3d1ec8b1216e4deba98e50160f4c480a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR