1-(4-Bromophenyl)piperidine ≥98.0%

Supplier: TCI America

B3930-1G B3930-5G
TCB3930-1GEA 74.26 CAD
TCB3930-1G TCB3930-5G
1-(4-Bromophenyl)piperidine ≥98.0%
1-(4-Bromophenyl)piperidine

CAS Number: 22148-20-5
MDL Number: MFCD04112533
Molecular Formula: C11H14BrN
Molecular Weight: 240.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 76

Formula: C₁₁H₁₄BrN
MW: 240.14 g/mol
Melting Pt: 76 °C
MDL Number: MFCD04112533
CAS Number: 22148-20-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc62e041f9610b984196a1b320f4101027ef918d&vid=cc62e041f9610b984196a1b320f4101027ef918d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR