3-Bromo-2-fluorobenzoic acid ≥98.0%

Supplier: TCI America

B3929-1G B3929-5G
TCB3929-1GEA 60.04 CAD
TCB3929-1G TCB3929-5G
3-Bromo-2-fluorobenzoic acid ≥98.0%
3-Bromo-2-fluorobenzoic acid

CAS Number: 161957-56-8
MDL Number: MFCD00665763
Molecular Formula: C7H4BrFO2
Molecular Weight: 219.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 170

Formula: C₇H₄BrFO₂
Melting Pt: 170 °C
MDL Number: MFCD00665763
CAS Number: 161957-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=310e12bdde6f2cfd77aac5d97a29d1234ed42220&vid=310e12bdde6f2cfd77aac5d97a29d1234ed42220&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR